Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCKBR Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5145222
Description
Immunogen sequence: FMHRRFRQAC LETCARCCPR PPRARPRALP DEDPPTPSIA SLSRLSYTTI STLGP Antigen sequence identity: Mouse 96%; Rat 95%.
CCKBR is a G-protein coupled receptor for gastrin and cholecystokinin (CCK), regulatory peptides of the brain and gastrointestinal tract. This protein is a type B gastrin receptor, which has a high affinity for both sulfated and nonsulfated CCK analogs and is found principally in the central nervous system and the gastrointestinal tract. A misspliced transcript variant including an intron has been observed in cells from colorectal and pancreatic tumors. CCKBR has been reported in brain, colon, pancreas, stomach, thyroid, and various cancers.
Specifications
| CCKBR | |
| Polyclonal | |
| Unconjugated | |
| CCKBR | |
| CCK(B) receptor; CCK2 receptor; Cck2r; CCK2-R; CCKB; CCK-B; Cckb receptor; CCK-B receptor; Cckbr; CCK-BR; CCKB-R; CCKR-2; CCKRB; cholecystokinin B receptor; cholecystokinin-2 receptor; Cholecystokinin-B; Cholrec; GASR; gastrin receptor; gastrin/cholecystokinin type B receptor; HGNC:1571 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 887 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P32239 | |
| CCKBR | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 392-446. Recombinant protein control fragment (Product #RP-110240). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction