Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCKBR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518384
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518384 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518384 Supplier Invitrogen™ Supplier No. PA518384
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine and mouse based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

CCKBR is a G-protein coupled receptor for gastrin and cholecystokinin (CCK), regulatory peptides of the brain and gastrointestinal tract. This protein is a type B gastrin receptor, which has a high affinity for both sulfated and nonsulfated CCK analogs and is found principally in the central nervous system and the gastrointestinal tract. A misspliced transcript variant including an intron has been observed in cells from colorectal and pancreatic tumors. CCKBR has been reported in brain, colon, pancreas, stomach, thyroid, and various cancers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCKBR
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene CCKBR
Gene Accession No. P32239, P56481
Gene Alias CCK(B) receptor; CCK2 receptor; Cck2r; CCK2-R; CCKB; CCK-B; Cckb receptor; CCK-B receptor; Cckbr; CCK-BR; CCKB-R; CCKR-2; CCKRB; cholecystokinin B receptor; cholecystokinin-2 receptor; Cholecystokinin-B; Cholrec; GASR; gastrin receptor; gastrin/cholecystokinin type B receptor; HGNC:1571
Gene Symbols CCKBR
Host Species Goat
Immunogen Synthetic peptide sequence (FDGDSDSDSQSRVRNQ) corresponding to the internal amino acids of CCKBR (aa 251-266).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12426, 887
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.