Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCKBR Polyclonal Antibody, Invitrogen™

Catalog No. PIPA527169
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA527169 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA527169 Supplier Thermo Scientific Supplier No. PA527169

Rabbit Polyclonal Antibody

Recommended positive controls: Unboiled HeLa cell extract. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

CCKBR is a G-protein coupled receptor for gastrin and cholecystokinin (CCK), regulatory peptides of the brain and gastrointestinal tract. This protein is a type B gastrin receptor, which has a high affinity for both sulfated and nonsulfated CCK analogs and is found principally in the central nervous system and the gastrointestinal tract. A misspliced transcript variant including an intron has been observed in cells from colorectal and pancreatic tumors. CCKBR has been reported in brain, colon, pancreas, stomach, thyroid, and various cancers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCKBR
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene CCKBR
Gene Accession No. P32239
Gene Alias CCK(B) receptor; CCK2 receptor; Cck2r; CCK2-R; CCKB; CCK-B; Cckb receptor; CCK-B receptor; Cckbr; CCK-BR; CCKB-R; CCKR-2; CCKRB; cholecystokinin B receptor; cholecystokinin-2 receptor; Cholecystokinin-B; Cholrec; GASR; gastrin receptor; gastrin/cholecystokinin type B receptor; HGNC:1571
Gene Symbols CCKBR
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a region within amino acids 271 and 363 of Human CCK-BR
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 887
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.