Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL1 Monoclonal Antibody (01), Invitrogen™

Mouse Monoclonal Antibody
$285.50 - $493.00
Specifications
| Antigen | CCL1 |
|---|---|
| Clone | 01 |
| Concentration | 1 mg/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
PIMA543522
|
Invitrogen™
MA543522 |
50 μL |
Each for $285.50
|
|
|||||
PIMA542970
![]() |
Invitrogen™
MA542970 |
100 μL |
Each for $493.00
|
|
|||||
Description
Specificity: Human I-309/CCL1
This gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine is secreted by activated T cells and displays chemotactic activity for monocytes but not for neutrophils. It binds to the chemokine receptor CCR8.Specifications
| CCL1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative | |
| BF534335; C-C motif chemokine 1; C-C motif chemokine ligand 1; CCL 1; Ccl1; CCR8 ligand; chemokine (C-C motif) ligand 1; I-309; inflammatory cytokine I-309; P500; SCYA1; SISe; SIS-epsilon; small inducible cytokine A1; small inducible cytokine A1 (I-309, homologous to mouse Tca-3); small-inducible cytokine A1; T lymphocyte-secreted protein I-309; TCA3; TCA-3; T-cell activation protein 3 | |
| CCL1 | |
| IgG2a | |
| Protein A | |
| Antibody |
| 01 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| P22362 | |
| 6346 | |
| Recombinant Human I-309/CCL1 Protein. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| CCL1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title