Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL11/Eotaxin Antibody (OTI2H9), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271277AF594
Description
CCL11/Eotaxin Monoclonal specifically detects CCL11/Eotaxin in Human samples. It is validated for Western Blot.Specifications
CCL11/Eotaxin | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
CCL11 | |
Recombinant protein fragment corresponding to amino acids 24-97 of human CCL11/Eotaxin (NP_002977) produced in E.coli. | |
Primary | |
Human | |
IgG1 |
Western Blot | |
OTI2H9 | |
Western Blot | |
C-C motif chemokine 11, chemokine (C-C motif) ligand 11, Eosinophil chemotactic protein, eotaxin, eotaxin-1, MGC22554, SCYA11Small-inducible cytokine A11, small inducible cytokine subfamily A (Cys-Cys), member 11 (eotaxin) | |
Mouse | |
0.1 mL | |
6356 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction