Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL12/MCP-5 Antibody (RM0169-8J25) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$206.50
Specifications
Antigen | CCL12/MCP-5 |
---|---|
Clone | RM0169-8J25 |
Concentration | LYOPH |
Applications | Western Blot, Blocking Assay |
Classification | Monoclonal |
Description
CCL12/MCP-5 Monoclonal specifically detects CCL12/MCP-5 in Mouse samples. It is validated for Western Blot, Block/Neutralize.Specifications
CCL12/MCP-5 | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
Q62401.1 | |
20293 | |
IgG2 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
RM0169-8J25 | |
Western Blot, Blocking Assay | |
Unconjugated | |
Rat | |
Mouse | |
C-C motif chemokine 12, chemokine (C-C motif) ligand 12, MCP-5, Scya12 | |
Ccl12 | |
Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title