Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL14/HCC-1/HCC-3 Antibody (015), Alexa Fluor™ 405 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289521AF405
Description
CCL14/HCC-1/HCC-3 Monoclonal antibody specifically detects CCL14/HCC-1/HCC-3 in Human samples. It is validated for ELISASpecifications
CCL14/HCC-1/HCC-3 | |
Monoclonal | |
Alexa Fluor 405 | |
C-C motif chemokine 14, chemokine (C-C motif) ligand 14, Chemokine CC-1/CC-3, chemokine CC-3, CKB1, FLJ16015, HCC-1, HCC-1(1-74), HCC-1/HCC-3, HCC-3, hemofiltrate CC chemokine 1, MCIF, member 14, NCC2CC-1, NCC-2CKb1, new CC chemokine 2, SCYA14CC-3, Small-inducible cytokine A14 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human CCL14/HCC-1/HCC-3 (Q16627-1; Thr20-Asn93). | |
0.1 mL | |
Biologically Active Proteins, Cell Cycle and Replication, Chemokines and Cytokines | |
6358 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
015 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction