Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL16/HCC-4/LEC Mouse anti-Human, Alexa Fluor 594, Clone: 70218, R&D Systems™

Catalog No. IC328T100UG
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC328T100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC328T100UG Supplier R&D Systems Supplier No. IC328T100UG
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CCL16/HCC-4/LEC Monoclonal specifically detects CCL16/HCC-4/LEC in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen CCL16/HCC-4/LEC
Applications Flow Cytometry
Classification Monoclonal
Clone 70218
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. O15467
Gene Alias C-C motif chemokine 16, chemokine (C-C motif) ligand 16, Chemokine CC-4, Chemokine LEC, CKb12, HCC4, HCC-4, HCC-4Liver-expressed chemokine, LCC-1Lymphocyte and monocyte chemoattractant, LEC, liver CC chemokine-1, LMC, LMCSmall-inducible cytokine A16, monotactin-1, MTN-1, NCC-4, NCC4IL-10-inducible chemokine, NCC-4ILINCK, new CC chemokine 4, SCYA16MGC117051, SCYL4, small inducible cytokine subfamily A (Cys-Cys), member 16
Gene Symbols CCL16
Host Species Mouse
Immunogen E. coli-derived recombinant human CCL16/HCC-4 Gln24-Gln120 Accession # O15467
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6360
Test Specificity Detects human CCL16/HCC-4 in direct ELISAs and Western blots.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.