Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL16/HCC-4/LEC Antibody (70218), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB328L
This item is not returnable.
View return policy
Description
CCL16/HCC-4/LEC Monoclonal antibody specifically detects CCL16/HCC-4/LEC in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, ELISA (Matched Antibody Pair)Specifications
CCL16/HCC-4/LEC | |
Monoclonal | |
DyLight 550 | |
C-C motif chemokine 16, chemokine (C-C motif) ligand 16, Chemokine CC-4, Chemokine LEC, CKb12, HCC-4Liver-expressed chemokine, LCC-1Lymphocyte and monocyte chemoattractant, LEC, liver CC chemokine-1, LMCSmall-inducible cytokine A16, monotactin-1, MTN-1, NCC4IL-10-inducible chemokine, NCC-4ILINCK, new CC chemokine 4, SCYA16MGC117051, SCYL4, small inducible cytokine subfamily A (Cys-Cys), member 16 | |
E. coli-derived recombinant human CCL16/HCC-4, Gln24-Gln120, Accession # O15467 | |
0.5 mL | |
Biologically Active Proteins, Chemokines and Cytokines | |
6360 | |
Store at 4°C in the dark. | |
IgG1 |
Western Blot, Flow Cytometry, ELISA | |
70218 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction