Learn More
Invitrogen™ CCL16 Monoclonal Antibody, PeproTech®
Mouse Monoclonal Antibody
Supplier: Invitrogen™ 500M43500UG

Description
AA Sequence of recombinant protein: QPKVPEWVNT PSTCCLKYYE KVLPRRLVVG YRKALNCHLP AIIFVTKRNR EVCTNPNDDW VQEYIKDPNL PLLPTRNLST VKIITAKNGQ PQLLNSQ. Preparation: Produced in BALB/c mice immunized with highly pure Recombinant Human LEC (CCL16). Anti-Human LEC (CCL16)-specific antibody was purified from cell culture by Protein G affinity chromatography.
CCL16 gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. CCL16 displays chemotactic activity for lymphocytes and monocytes but not for neutrophils. This cytokine also shows a potent myelosuppressive activity and suppresses proliferation of myeloid progenitor cells. The expression of this gene is upregulated by IL-10.
Specifications
CCL16 | |
Monoclonal | |
PBS with no preservative | |
O15467 | |
CCL16 | |
E.coli-derived Recombinant Human LEC (CCL16). | |
500 μg | |
Primary | |
Human | |
Antibody |
ELISA, Western Blot | |
Unconjugated | |
CCL16 | |
C-C motif chemokine 16; C-C motif chemokine ligand 16; CCL16; chemokine (C-C motif) ligand 16; Chemokine CC-4; Chemokine LEC; CKb12; HCC-4; IL-10-inducible chemokine; ILINCK; LCC-1; LEC; liver CC chemokine-1; liver-expressed chemokine; LMC; Lymphocyte and monocyte chemoattractant; MGC117051; monotactin-1; MTN-1; NCC4; NCC-4; new CC chemokine 4; SCYA16; SCYL4; small inducible cytokine subfamily A (Cys-Cys), member 16; small-inducible cytokine A16 | |
Mouse | |
Protein G | |
RUO | |
6360 | |
-20°C | |
Lyophilized |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.