Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL17/TARC Antibody (54015), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB3641N
Description
CCL17/TARC Monoclonal antibody specifically detects CCL17/TARC in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
CCL17/TARC | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
54015 | |
Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
ABCD-2, CC chemokine TARC, C-C motif chemokine 17, chemokine (C-C motif) ligand 17, MGC138273, SCYA17MGC138271, small inducible cytokine subfamily A (Cys-Cys), member 17, Small-inducible cytokine A17, T cell-directed CC chemokine, TARCA-152E5.3, Thymus and activation-regulated chemokine | |
E. coli-derived recombinant human CCL17/TARC, Ala24-Ser94, Accession # Q92583.1 | |
0.5 mL | |
Biologically Active Proteins, Chemokines and Cytokines | |
6361 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction