Learn More
Description
Specifications
Specifications
| Antigen | CCL17/TARC |
| Applications | ELISA, Sandwich ELISA |
| Classification | Monoclonal |
| Conjugate | HRP |
| Dilution | ELISA, Sandwich ELISA |
| Gene Alias | ABCD-2, CC chemokine TARC, C-C motif chemokine 17, chemokine (C-C motif) ligand 17, MGC138273, SCYA17MGC138271, small inducible cytokine subfamily A (Cys-Cys), member 17, Small-inducible cytokine A17, T cell-directed CC chemokine, TARCA-152E5.3, Thymus and activation-regulated chemokine |
| Host Species | Mouse |
| Quantity | 5 Plates |
| Regulatory Status | RUO |
| Research Discipline | Biologically Active Proteins, Chemokines and Cytokines |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
