Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL17/TARC Antibody Pair, HRP, R&D Systems™
SDP

Catalog No. NB292581 Shop All Novus Products
Change view
Click to view available options
Quantity:
15 Plates
5 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292581 15 Plates
NB292580 5 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB292581 Supplier R&D Systems Supplier No. NBP32610915PLATES
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

CCL17/TARC Monoclonal specifically detects CCL17/TARC in Human samples. It is validated for ELISA∣Sandwich ELISA.

Specifications

Antigen CCL17/TARC
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Conjugate HRP
Dilution ELISA, Sandwich ELISA
Gene Alias ABCD-2, CC chemokine TARC, C-C motif chemokine 17, chemokine (C-C motif) ligand 17, MGC138273, SCYA17MGC138271, small inducible cytokine subfamily A (Cys-Cys), member 17, Small-inducible cytokine A17, T cell-directed CC chemokine, TARCA-152E5.3, Thymus and activation-regulated chemokine
Host Species Mouse
Quantity 15 Plates
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Chemokines and Cytokines
Primary or Secondary Primary
Gene ID (Entrez) 6361
Target Species Human
Content And Storage Storage of components varies. See protocol for specific instructions.
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.