Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL19/MIP-3 beta Antibody (MM0143-11B4), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212097F
Description
CCL19/MIP-3 beta Monoclonal specifically detects CCL19/MIP-3 beta in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
CCL19/MIP-3 beta | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
CCL19 | |
Human recombinant CCL19/MIP-3 beta | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
MM0143-11B4 | |
Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
beta chemokine exodus-3, Beta-chemokine exodus-3, CC chemokine ligand 19, C-C motif chemokine 19, chemokine (C-C motif) ligand 19, CKb11, EBI1-ligand chemokine, ELCMIP-3-beta, Epstein-Barr virus-induced molecule 1 ligand chemokine, exodus-3, Macrophage inflammatory protein 3 beta, macrophage inflammatory protein 3-beta, MGC34433, MIP-3b, MIP3BCK beta-11, SCYA19EBI1 ligand chemokine, small inducible cytokine subfamily A (Cys-Cys), member 19, Small-inducible cytokine A19 | |
Mouse | |
Protein G purified | |
Biologically Active Proteins | |
6363 | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction