Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL19/MIP-3 beta Antibody (SR1080), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | CCL19/MIP-3 beta |
---|---|
Clone | SR1080 |
Dilution | Western Blot 1:1000-1:5000, Flow Cytometry 1:100 |
Applications | Western Blot, Flow Cytometry |
Classification | Monoclonal |
Description
CCL19/MIP-3 beta Monoclonal antibody specifically detects CCL19/MIP-3 beta in Human samples. It is validated for Western Blot, Flow CytometrySpecifications
CCL19/MIP-3 beta | |
Western Blot 1:1000-1:5000, Flow Cytometry 1:100 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
beta chemokine exodus-3, Beta-chemokine exodus-3, CC chemokine ligand 19, C-C motif chemokine 19, chemokine (C-C motif) ligand 19, CKb11, EBI1-ligand chemokine, ELCMIP-3-beta, Epstein-Barr virus-induced molecule 1 ligand chemokine, exodus-3, Macrophage inflammatory protein 3 beta, macrophage inflammatory protein 3-beta, MGC34433, MIP-3b, MIP3BCK beta-11, SCYA19EBI1 ligand chemokine, small inducible cytokine subfamily A (Cys-Cys), member 19, Small-inducible cytokine A19 | |
A synthesized peptide derived from human CCL19/MIP-3 beta (Uniprot #: Q99731) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1080 | |
Western Blot, Flow Cytometry | |
Unconjugated | |
Rabbit | |
Biologically Active Proteins | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
6363 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title