Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL19/MIP-3 beta Antibody (RM0171-10J38) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$206.50
Specifications
Antigen | CCL19/MIP-3 beta |
---|---|
Clone | RM0171-10J38 |
Concentration | LYOPH |
Applications | Western Blot, Blocking Assay |
Classification | Monoclonal |
Description
CCL19/MIP-3 beta Monoclonal specifically detects CCL19/MIP-3 beta in Mouse samples. It is validated for Western Blot, Block/Neutralize.Specifications
CCL19/MIP-3 beta | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
Q548P0 | |
6363 | |
IgG2 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
RM0171-10J38 | |
Western Blot, Blocking Assay | |
Unconjugated | |
Rat | |
Mouse | |
beta chemokine exodus-3, Beta-chemokine exodus-3, CC chemokine ligand 19, C-C motif chemokine 19, chemokine (C-C motif) ligand 19, CKb11, EBI1-ligand chemokine, ELCMIP-3-beta, Epstein-Barr virus-induced molecule 1 ligand chemokine, exodus-3, Macrophage inflammatory protein 3 beta, macrophage inflammatory protein 3-beta, MGC34433, MIP-3b, MIP3BCK beta-11, SCYA19EBI1 ligand chemokine, small inducible cytokine subfamily A (Cys-Cys), member 19, Small-inducible cytokine A19 | |
CCL19 | |
Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title