Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL19 Mouse anti-Human, Clone: OTI4F3, lyophilized, TrueMAB™
SDP

Catalog No. 501666983
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-6983 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-6983 Supplier OriGene Supplier No. CF804426
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

Macrophage Inflammatory Protein-3 beta (MIP-3 beta), also called CCL19, is a chemokine that is expressed in the thymus, lymph nodes and activated bone marrow stromal cells. MIP-3 beta signals through the G protein-coupled receptor, CCR7, to regulate normal lymphocyte recirculation and homing of T and B cells to the lymph nodes. Human MIP-3 beta shows activity on mouse cells.

Specifications

Antigen CCL19
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI4F3
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene CCL19
Gene Accession No. Q99731
Gene Alias CKb11, ELC, MIP-3b, MIP3B, SCYA19
Gene Symbols CCL19
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 22-98 of human CCL19 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6363
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.