Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL2/JE/MCP-1 Antibody (001), PerCP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290550PCP
Description
CCL2/JE/MCP-1 Monoclonal antibody specifically detects CCL2/JE/MCP-1 in Mouse samples. It is validated for ELISA, ELISASpecifications
CCL2/JE/MCP-1 | |
Monoclonal | |
PerCP | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
6347 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
001 | |
ELISA, Sandwich ELISA Detection | |
C-C motif chemokine ligand 2, CCL2, GDCF-2, HC11, HSMCR30, MCAF, Mcp1, Mcp-1, SCYA2, SMC-CF | |
Rabbit immunized with purified, recombinant Mouse CCL2/JE/MCP-1 (Accession#: NP_035463.1, Met1-Asn148) | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction