Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL2/JE/MCP-1 Antibody (123616R), DyLight 755, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB479RZ
Description
CCL2/JE/MCP-1 Monoclonal antibody specifically detects CCL2/JE/MCP-1 in Mouse samples. It is validated for Flow CytometrySpecifications
CCL2/JE/MCP-1 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from cell culture supernatant | |
RUO | |
6347 | |
Store at 4°C in the dark. | |
IgG2b |
Flow Cytometry | |
123616R | |
Flow Cytometry | |
C-C motif chemokine ligand 2, CCL2, GDCF-2, HC11, HSMCR30, MCAF, Mcp1, Mcp-1, SCYA2, SMC-CF | |
E. coli-derived recombinant mouse CCL2, Gln24-Arg96, Accession # P10148 | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction