Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL2/JE/MCP-1 Antibody (144), HRP

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289290H
Description
CCL2/JE/MCP-1 Monoclonal specifically detects CCL2/JE/MCP-1 in Human samples. It is validated for ELISA, Sandwich ELISA Detection.Specifications
CCL2/JE/MCP-1 | |
Monoclonal | |
HRP | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
6347 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
144 | |
ELISA, Sandwich ELISA Detection | |
C-C motif chemokine ligand 2, CCL2, GDCF-2, HC11, HSMCR30, MCAF, Mcp1, Mcp-1, SCYA2, SMC-CF | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human CCL2/JE/MCP-1 (NP_002973.1; Gln24-Thr99). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction