Learn More
CCL2 (MCP-1) Monoclonal Antibody (2H5), Functional Grade, eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
$3260.00
Specifications
| Antigen | CCL2 (MCP-1) |
|---|---|
| Clone | 2H5 |
| Concentration | 1 mg/mL |
| Applications | Functional Assay, Neutralization |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
16-709-638
|
Invitrogen™
16709638 |
5 mg |
Each of 1 for $3,260.00
|
|
|||||
Description
The 2H5 MAb reacts with mouse, rat, and human monocyte chemoattractant protein-1 (MCP-1), also known as CCL2 and MCAF.
This gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine displays chemotactic activity for monocytes and basophils but not for neutrophils or eosinophils. It has been implicated in the pathogenesis of diseases characterized by monocytic infiltrates, like psoriasis, rheumatoid arthritis and atherosclerosis. It binds to chemokine receptors CCR2 and CCR4.Specifications
| CCL2 (MCP-1) | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.2 | |
| 20296, 24770, 6347 | |
| Primary | |
| 4°C | |
| Ccl2 |
| 2H5 | |
| Functional Assay, Neutralization | |
| Functional Grade | |
| Armenian Hamster | |
| Human, Mouse, Rat | |
| P10148, P13500, P14844 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.