Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL2/MCP1 Antibody (ECE.2), Novus Biologicals™

Rat Monoclonal Antibody has been used in 4 publications
Supplier: Novus Biologicals NBP142312
Description
CCL2/MCP1 Monoclonal specifically detects CCL2/MCP1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen, Immunoassay.Specifications
CCL2/MCP1 | |
Monoclonal | |
0.1 mg/mL | |
Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen, Immunoassay | |
CCL2 | |
The antibody reacts with murine JE MCP-1. | |
0.2 mL | |
Apoptosis, Cancer, Chemokine Research, Cytokine Research, Hypoxia, Immunology, Innate Immunity, Signal Transduction | |
6347 | |
Store at 4C. Do not freeze. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoassay | |
ECE.2 | |
Unconjugated | |
C-C motif chemokine 2, chemokine (C-C motif) ligand 2, GDCF-2, HC11Monocyte secretory protein JE, MCAFMonocyte chemotactic and activating factor, MCP1 Monocyte chemotactic protein 1, MCP-1 Small-inducible cytokine A2, MGC9434, Monocyte chemoattractant protein 1, monocyte chemoattractant protein-1, SCYA2HSMCR30, small inducible cytokine A2 (monocyte chemotactic protein 1, homologous tomouse Sig-je), small inducible cytokine subfamily A (Cys-Cys), member 2, SMC-CF | |
Rat | |
Protein A or G purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction