Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Ccl2 (Mouse) ELISA Kit
Shop All Abnova Corporation Products
Click to view available options
Quantity:
1 Kit
Description
Ccl2 (Mouse) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for the quantitative measurement of mouse Ccl2.
Specifications
Specifications
Cross Reactivity | Mouse |
For Use With (Application) | Quantitative |
Gene | Ccl2 |
Gene Alias | AI323594, HC11, JE, MCAF, MCP-1, MCP1, SMC-CF, Scya2, Sigje |
Gene ID (Entrez) | 20296 |
Quantity | 1 Kit |
Regulatory Status | RUO |
Storage Requirements | Store at 4°C for 6 months, at -20°C for 12 months. Avoid multiple freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction