Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL20/MIP-3 alpha Antibody (206D9.05), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals DDX0420A488100
Description
CCL20/MIP-3 alpha Monoclonal specifically detects CCL20/MIP-3 alpha in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
CCL20/MIP-3 alpha | |
Monoclonal | |
0.5mg/mL | |
Western Blot, Immunohistochemistry | |
CCL20 | |
MIP-3 Alpha/CCL20 was used as immunogen. | |
0.1 mg | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry | |
206D9.05 | |
Alexa Fluor 488 | |
beta chemokine exodus-1, Beta-chemokine exodus-1, CC chemokine LARC, C-C motif chemokine 20, chemokine (C-C motif) ligand 20, CKb4, exodus-1, LARCLiver and activation-regulated chemokine, MIP-3a, MIP-3-alpha, MIP3AMacrophage inflammatory protein 3 alpha, SCYA20Small-inducible cytokine A20, small inducible cytokine subfamily A (Cys-Cys), member 20, ST38 | |
Mouse | |
Ion exchange chromatography | |
RUO | |
6364 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction