Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL20/MIP-3 alpha Antibody (4N8C7), DyLight 680, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP227143FR
Description
CCL20/MIP-3 alpha Monoclonal antibody specifically detects CCL20/MIP-3 alpha in Mouse samples. It is validated for Western Blot.Specifications
CCL20/MIP-3 alpha | |
Monoclonal | |
DyLight 680 | |
beta chemokine exodus-1, Beta-chemokine exodus-1, CC chemokine LARC, C-C motif chemokine 20, chemokine (C-C motif) ligand 20, CKb4, exodus-1, LARCLiver and activation-regulated chemokine, MIP-3a, MIP-3-alpha, MIP3AMacrophage inflammatory protein 3 alpha, SCYA20Small-inducible cytokine A20, small inducible cytokine subfamily A (Cys-Cys), member 20, ST38 | |
Rat | |
Protein G purified | |
RUO | |
6364 | |
Mouse | |
Purified |
Western Blot | |
4N8C7 | |
Western Blot | |
CCL20 | |
Full length recombinant mouse CCL20 (MIP-3a) was used as immunogen for the antibody. full length protein | |
0.1 mL | |
Primary | |
This monoclonal antibody is specific to Mouse CCL20, it will not cross-react with Human CCL20. | |
Store at 4C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction