Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL20/MIP-3 alpha Antibody (4N8C7), Novus Biologicals™

Rat Monoclonal Antibody
$206.50 - $426.00
Specifications
Antigen | CCL20/MIP-3 alpha |
---|---|
Clone | 4N8C7 |
Concentration | 1.0 mg/ml |
Dilution | Western Blot 0.1-2ug/ml |
Applications | Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP227143SS
![]() |
Novus Biologicals
NBP227143SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP227143
![]() |
Novus Biologicals
NBP227143 |
0.1 mg |
Each for $426.00
|
|
|||||
Description
CCL20/MIP-3 alpha Monoclonal specifically detects CCL20/MIP-3 alpha in Mouse samples. It is validated for Western Blot.Specifications
CCL20/MIP-3 alpha | |
1.0 mg/ml | |
Western Blot | |
Unconjugated | |
Rat | |
Chemokines and Cytokines | |
O89093 | |
6364 | |
Full length recombinant mouse CCL20 (MIP-3a) was used as immunogen for the antibody. full length protein | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
4N8C7 | |
Western Blot 0.1-2ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
beta chemokine exodus-1, Beta-chemokine exodus-1, CC chemokine LARC, C-C motif chemokine 20, chemokine (C-C motif) ligand 20, CKb4, exodus-1, LARCLiver and activation-regulated chemokine, MIP-3a, MIP-3-alpha, MIP3AMacrophage inflammatory protein 3 alpha, SCYA20Small-inducible cytokine A20, small inducible cytokine subfamily A (Cys-Cys), member 20, ST38 | |
CCL20 | |
IgG2a κ | |
Protein G purified | |
This monoclonal antibody is specific to Mouse CCL20, it will not cross-react with Human CCL20. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title