Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL20/MIP-3 alpha Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP145246
Description
CCL20/MIP-3 alpha Polyclonal specifically detects CCL20/MIP-3 alpha in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
CCL20/MIP-3 alpha | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
beta chemokine exodus-1, Beta-chemokine exodus-1, CC chemokine LARC, C-C motif chemokine 20, chemokine (C-C motif) ligand 20, CKb4, exodus-1, LARCLiver and activation-regulated chemokine, MIP-3a, MIP-3-alpha, MIP3AMacrophage inflammatory protein 3 alpha, SCYA20Small-inducible cytokine A20, small inducible cytokine subfamily A (Cys-Cys), member 20, ST38 | |
Goat | |
Affinity Purified | |
RUO | |
6364 | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
0.5 mg/mL | |
Immunohistochemistry 6 - 8 ug/ml, Immunohistochemistry-Paraffin 6 - 8 ug/ml, Peptide ELISA Detection limit 1:16000 | |
P78556 | |
CCL20 | |
Peptide with sequence CLGYTDRILHPK corresponding to internal region according to NP_004582.1, NP_001123518.1. | |
0.1 mg | |
Primary | |
This antibody is expected to recognize both isoforms (NP_004582.1; NP_001123518.1). | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction