Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL20/MIP-3 alpha Antibody (4N5F7) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP280619
Description
CCL20/MIP-3 alpha Monoclonal specifically detects CCL20/MIP-3 alpha in Human samples. It is validated for Western Blot.Specifications
CCL20/MIP-3 alpha | |
Monoclonal | |
1 mg/mL | |
Western Blot 0.1-2ug/ml | |
beta chemokine exodus-1, Beta-chemokine exodus-1, CC chemokine LARC, C-C motif chemokine 20, chemokine (C-C motif) ligand 20, CKb4, exodus-1, LARCLiver and activation-regulated chemokine, MIP-3a, MIP-3-alpha, MIP3AMacrophage inflammatory protein 3 alpha, SCYA20Small-inducible cytokine A20, small inducible cytokine subfamily A (Cys-Cys), member 20, ST38 | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
This monoclonal antibody is specific to mouse MIP-3 it will not cross-react with human MIP-3 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG2a κ |
Western Blot | |
4N5F7 | |
Unconjugated | |
PBS | |
CCL20 | |
Full length recombinant mouse MIP-3 full length protein | |
0.1 mg | |
Biologically Active Proteins, Chemokines and Cytokines | |
6364 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction