Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCL20 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595917
Description
Positive Samples: K-562, LO2, A-549, BxPC-3 Immunogen sequence: MCCTKSLLLA ALMSVLLLHL CGESEAASNF DCCLGYTDRI LHPKFIVGFT RQLANEGCDI NAIIFHTKKK LSVCANPKQT WVKYIVRLLS KKVKN.
Macrophage Inflammatory Protein-3 alpha (MIP-3alpha), also called CCL20, is expressed in the liver, lungs, lymph nodes and peripheral blood lymphocytes. MIP-3alpha is strongly up regulated by inflammatory signals, and down regulated by the anti-inflammatory cytokine IL-10. MIP3alpha signals through the G protein-coupled receptor, CCR6, and acts as a chemoattractant to lymphocytes and dendritic cells.
Specifications
CCL20 | |
Polyclonal | |
Unconjugated | |
CCL20 | |
beta chemokine exodus-1; beta-chemokine exodus-1; CC chemokine LARC; CC chemokine ST38; C-C motif chemokine 20; C-C motif chemokine ligand 20; Ccl20; CCL20(1-64); CCL20(1-67); CCL20(2-70); chemokine (C-C motif) ligand 20; CKb4; Exodus; exodus-1; LARC; Liver and activation-regulated chemokine; macrophage inflammatory protein 3 alpha; MIP 3 a; MIP 3 a; MIP 3alpha; MIP 3a; MIP3 a; MIP3 alpha; MIP3 a; MIP-3[a]; MIP3A; MIP-3A; MIP-3-alpha; M-MIP-3-alpha; SCYA20; small inducible cytokine subfamily A (Cys-Cys), member 20; small inducible cytokine subfamily A20; small-inducible cytokine A20; St38 | |
Rabbit | |
Affinity chromatography | |
RUO | |
6364 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.72 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P78556 | |
CCL20 | |
A synthetic peptide corresponding to a sequence within amino acids 1-95 of human CCL20 (NP_0045821). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction