Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL21/6Ckine Antibody (MM0146-2H26), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212103MFV500
Description
CCL21/6Ckine Monoclonal antibody specifically detects CCL21/6Ckine in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ImmunocytochemistrySpecifications
CCL21/6Ckine | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry | |
MM0146-2H26 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunocytochemistry | |
6CkineSmall-inducible cytokine A21, Beta-chemokine exodus-2, chemokine (C-C motif) ligand 21, CKb9, member 21, SCYA21MGC34555, secondary lymphoid tissue chemokine, SLCSecondary lymphoid-tissue chemokine | |
Human recombinant CCL21/6Ckine | |
0.1 mL | |
Biologically Active Proteins, Cancer, Cell Cycle and Replication, Chemokines and Cytokines | |
6366 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction