Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL21/6Ckine Antibody (SR1207), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322286100UL
Description
CCL21/6Ckine Monoclonal antibody specifically detects CCL21/6Ckine in Human samples. It is validated for Western BlotSpecifications
CCL21/6Ckine | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
SR1207 | |
Western Blot 1:500-1:2000 | |
6CkineSmall-inducible cytokine A21, Beta-chemokine exodus-2, chemokine (C-C motif) ligand 21, CKb9, member 21, SCYA21MGC34555, secondary lymphoid tissue chemokine, SLCSecondary lymphoid-tissue chemokine | |
A synthesized peptide derived from human CCL21/6Ckine (Uniprot #: O00585) | |
100 μg | |
Biologically Active Proteins, Cancer, Cell Cycle and Replication, Chemokines and Cytokines | |
6366 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction