Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCL21 Monoclonal Antibody (05)
GREENER_CHOICE

Catalog No. PIMA529848
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529848 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529848 Supplier Invitrogen™ Supplier No. MA529848
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Rhesus CCL21/6Ckine.

This gene is one of several CC cytokine genes clustered on the p-arm of chromosome 9. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. Similar to other chemokines the protein encoded by this gene inhibits hemopoiesis and stimulates chemotaxis. This protein is chemotactic in vitro for thymocytes and activated T cells, but not for B cells, macrophages, or neutrophils. The cytokine encoded by this gene may also play a role in mediating homing of lymphocytes to secondary lymphoid organs. It is a high affinity functional ligand for chemokine receptor 7 (CCR7) that is expressed on T and B lymphocytes and a known receptor for another member of the cytokine family (small inducible cytokine A19).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCL21
Applications ELISA
Classification Monoclonal
Clone 05
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CCL21
Gene Accession No. Q8HYP5
Gene Alias 6 Ckine; 6CKBAC1; 6CKBAC2; 6Ckine; AI159701; ALP; AW987545; beta chemokine exodus-2; beta-chemokine exodus-2; CC chemokine 6Ckine-ser; C-C motif chemokine 21; C-C motif chemokine 21a; C-C motif chemokine 21b; C-C motif chemokine 21c; C-C motif chemokine ligand 21; CCL21; Ccl21a; Ccl21b; Ccl21c; Ccl21-leu; CCL21-Ser; chemokine (C-C motif) ligand 21; chemokine (C-C motif) ligand 21A (serine); chemokine (C-C motif) ligand 21B (leucine); chemokine (C-C motif) ligand 21b (serine); chemokine (C-C motif) ligand 21C (leucine); chemokine CCL21/6CKINE; CKb9; ECL; Efficient Chemoattractant for Lymphocytes; exodus-2; plt; Scya21; SCYA21a; Scya21b; Scya21c; secondary lymphoid organ chemokine; secondary lymphoid tissue chemokine; Secondary lymphoid-tissue chemokine; SLC; small inducible cytokine A21; small inducible cytokine A21a; small inducible cytokine A21c; small inducible cytokine subfamily A (Cys-Cys), member 21; small-inducible cytokine A21; Small-inducible cytokine A21a; Small-inducible cytokine A21b; Small-inducible cytokine A21c; TCA4; Thymus-derived chemotactic agent 4; UNQ784/PRO1600
Gene Symbols CCL21
Host Species Mouse
Immunogen Recombinant Rhesus CCL21/6Ckine protein (Met1-Pro131).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 574183
Target Species Rhesus Macaque
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.