Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PeproTech CCL21 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Catalog No. 502718708
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μg
50 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-271-8708 500 μg
500P109BT25 25 μg
500P109BT50 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-271-8708 Supplier PeproTech Supplier No. 500P109BT500UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: discontinued. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human Exodus-2 (CCL21). Anti-Human Exodus-2 (CCL21)-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human Exodus-2 (CCL21) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human Exodus-2 (CCL21) (500-P109) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human Exodus-2 (CCL21). Western Blot: To detect hExodus-2 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hExodus-2 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P109BT-1MG will be provided as 2 x 500 μg

This gene is one of several CC cytokine genes clustered on the p-arm of chromosome 9. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. Similar to other chemokines the protein encoded by this gene inhibits hemopoiesis and stimulates chemotaxis. This protein is chemotactic in vitro for thymocytes and activated T cells, but not for B cells, macrophages, or neutrophils. The cytokine encoded by this gene may also play a role in mediating homing of lymphocytes to secondary lymphoid organs. It is a high affinity functional ligand for chemokine receptor 7 (CCR7) that is expressed on T and B lymphocytes and a known receptor for another member of the cytokine family (small inducible cytokine A19).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCL21
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Biotin
Formulation PBS with no preservative
Gene CCL21
Gene Accession No. O00585
Gene Alias 6 Ckine; 6CKBAC1; 6CKBAC2; 6Ckine; AI159701; ALP; AW987545; beta chemokine exodus-2; beta-chemokine exodus-2; CC chemokine 6Ckine-ser; C-C motif chemokine 21; C-C motif chemokine 21a; C-C motif chemokine 21b; C-C motif chemokine 21c; C-C motif chemokine ligand 21; CCL21; Ccl21a; Ccl21b; Ccl21c; Ccl21-leu; CCL21-Ser; chemokine (C-C motif) ligand 21; chemokine (C-C motif) ligand 21A (serine); chemokine (C-C motif) ligand 21B (leucine); chemokine (C-C motif) ligand 21b (serine); chemokine (C-C motif) ligand 21C (leucine); chemokine CCL21/6CKINE; CKb9; ECL; Efficient Chemoattractant for Lymphocytes; exodus-2; plt; Scya21; SCYA21a; Scya21b; Scya21c; secondary lymphoid organ chemokine; secondary lymphoid tissue chemokine; Secondary lymphoid-tissue chemokine; SLC; small inducible cytokine A21; small inducible cytokine A21a; small inducible cytokine A21c; small inducible cytokine subfamily A (Cys-Cys), member 21; small-inducible cytokine A21; Small-inducible cytokine A21a; Small-inducible cytokine A21b; Small-inducible cytokine A21c; TCA4; Thymus-derived chemotactic agent 4; UNQ784/PRO1600
Gene Symbols CCL21
Host Species Rabbit
Immunogen E.coli-derived Recombinant Human Exodus-2 (CCL21).
Purification Method Antigen affinity chromatography
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6366
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.