Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL22/MDC Antibody (RM0175-9L23), Alexa Fluor™ 488, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212106AF488
Description
CCL22/MDC Monoclonal specifically detects CCL22/MDC in Mouse samples. It is validated for Western Blot.Specifications
CCL22/MDC | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CCL22 | |
Mouse recombinant CCL22/MDC | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Western Blot | |
RM0175-9L23 | |
Western Blot | |
A-152E5.1, ABCD-1, CC chemokine STCP-1, C-C motif chemokine 22, chemokine (C-C motif) ligand 22, DC/B-CK, Macrophage-derived chemokine, MDCStimulated T-cell chemotactic protein 1, MGC34554, SCYA22MDC(1-69), small inducible cytokine A22, small inducible cytokine subfamily A (Cys-Cys), member 22, Small-inducible cytokine A22, STCP-1, stimulated T cell chemotactic protein 1 | |
Rat | |
Protein A or G purified | |
Biologically Active Proteins | |
6367 | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction