Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL23/MPIF-1 Mouse anti-Human, Alexa Fluor 647, Clone: 69512, R&D Systems™

Catalog No. IC371R100UG
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC371R100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC371R100UG Supplier R&D Systems Supplier No. IC371R100UG
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CCL23/MPIF-1 Monoclonal antibody specifically detects CCL23/MPIF-1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen CCL23/MPIF-1
Applications Flow Cytometry
Classification Monoclonal
Clone 69512
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 647
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P55773
Gene Symbols CCL23
Host Species Mouse
Immunogen E. coli-derived recombinant human CCL23/MPIF-1 Arg22-Asn120 Accession # P55773
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects human CCL23/MPIF-1 in direct ELISAs and Western blots.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.