Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL23/MPIF-1 Mouse anti-Human, Clone: 69512, R&D Systems™

Catalog No. MAB371SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB371SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB371SP Supplier R&D Systems Supplier No. MAB371SP
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CCL23/MPIF-1 Monoclonal antibody MAB371SP specifically detects CCL23/MPIF-1 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Neutralization, Western Blotting

Specifications

Antigen CCL23/MPIF-1
Applications Western Blot, Neutralization, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 69512
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 2-6 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P55773
Gene Symbols CCL23
Host Species Mouse
Immunogen E. coli-derived recombinant human CCL23/MPIF-1 Arg22-Asn120 Accession # P55773
Purification Method Protein A or G purified from ascites
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6368
Test Specificity Detects human CCL23/MPIF-1 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.