Learn More
Invitrogen™ CCL23 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA547528
Description
In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human (rh) MIP-1 gamma and rhHCC-1 is observed and 5% cross-reactivity with recombinant mouse MIP-1 gamma is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
This gene is one of several chemokine genes clustered on the q-arm of chromosome 17. Chemokines form a superfamily of secreted proteins involved in immunoregulatory and inflammatory processes. The superfamily is divided into four subfamilies based on the arrangement of the N-terminal cysteine residues of the mature peptide. This chemokine, a member of the CC subfamily, displays chemotactic activity on resting T lymphocytes and monocytes, lower activity on neutrophils and no activity on activated T lymphocytes. The protein is also a strong suppressor of colony formation by a multipotential hematopoietic progenitor cell line. In addition, the product of this gene is a potent agonist of the chemokine (C-C motif) receptor 1. Alternative splicing results in multiple transcript variants that encode different isoforms.
Specifications
| CCL23 | |
| Polyclonal | |
| Unconjugated | |
| CCL23 | |
| C6 beta-chemokine; C-C motif chemokine 23; C-C motif chemokine ligand 23; CCL23; CCL23(19-99); CCL23(22-99); CCL23(27-99); CCL23(30-99); chemokine (C-C motif) ligand 23; Ck beta 8-1; CKb8; CKB-8; Ckb-8-1; CK-BETA-8; hmrp-2a; Macrophage inflammatory protein 3; MIP3; MIP-3; MIP-3CK-BETA-8; MPIF1; MPIF-1; MPIF-1Ckb-8-1; Myeloid progenitor inhibitory factor 1; SCYA23; small inducible cytokine subfamily A (Cys-Cys), member 23; Small-inducible cytokine A23 | |
| Goat | |
| Antigen Affinity Chromatography | |
| RUO | |
| 6368 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Neutralization, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P55773 | |
| CCL23 | |
| E. coli-derived recombinant human CCL23/MPIF-1 Arg22-Asn120. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.