Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCL23 Recombinant Rabbit Monoclonal Antibody (007), Invitrogen™

Catalog No. PIMA543467
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543467 50 μL
PIMA543066 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543467 Supplier Invitrogen™ Supplier No. MA543467
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Specificity: Human CCL23/MIP 3

This gene is one of several chemokine genes clustered on the q-arm of chromosome 17. Chemokines form a superfamily of secreted proteins involved in immunoregulatory and inflammatory processes. The superfamily is divided into four subfamilies based on the arrangement of the N-terminal cysteine residues of the mature peptide. This chemokine, a member of the CC subfamily, displays chemotactic activity on resting T lymphocytes and monocytes, lower activity on neutrophils and no activity on activated T lymphocytes. The protein is also a strong suppressor of colony formation by a multipotential hematopoietic progenitor cell line. In addition, the product of this gene is a potent agonist of the chemokine (C-C motif) receptor 1. Alternative splicing results in multiple transcript variants that encode different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCL23
Applications ELISA
Classification Recombinant Monoclonal
Clone 007
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CCL23
Gene Accession No. P55773
Gene Alias C6 beta-chemokine; C-C motif chemokine 23; C-C motif chemokine ligand 23; CCL23; CCL23(19-99); CCL23(22-99); CCL23(27-99); CCL23(30-99); chemokine (C-C motif) ligand 23; Ck beta 8-1; CKb8; CKB-8; Ckb-8-1; CK-BETA-8; hmrp-2a; Macrophage inflammatory protein 3; MIP3; MIP-3; MIP-3CK-BETA-8; MPIF1; MPIF-1; MPIF-1Ckb-8-1; Myeloid progenitor inhibitory factor 1; SCYA23; small inducible cytokine subfamily A (Cys-Cys), member 23; Small-inducible cytokine A23
Gene Symbols CCL23
Host Species Rabbit
Immunogen Recombinant Human CCL23/MIP 3 Protein.
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6368
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.