Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL24/Eotaxin-2/MPIF-2 Antibody (61016.11) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB12010370
Description
CCL24/Eotaxin-2/MPIF-2 Monoclonal specifically detects CCL24/Eotaxin-2/MPIF-2 in Human samples. It is validated for Western Blot, ELISA.Specifications
CCL24/Eotaxin-2/MPIF-2 | |
Monoclonal | |
0.5 mg/mL | |
Western Blot 1-2 ug/ml, ELISA 1:100-1:2000 | |
CCL24 | |
Purified recombinant human eotaxin 2, expressed in E. coli as immunogen. | |
0.1 mg | |
Cytokine Research | |
6369 | |
Human | |
Purified |
Western Blot, ELISA | |
61016.11 | |
Unconjugated | |
chemokine (C-C motif) ligand 24, Ckb-6, Eosinophil chemotactic protein 2, Eotaxin-2, member 24, MPIF2Myeloid progenitor inhibitory factor 2, MPIF-2Small-inducible cytokine A24, SCYA24CK-beta-6 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
When used as a capture in human eotaxin 2 sandwich ELISAs, this exhibits less than 0.03 % cross-reactivity with recombinant human eotaxin, MIP1 alpha, and MCP3. | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction