Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL24/Eotaxin-2/MPIF-2 Antibody (61016.11), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB12010370UV
Description
CCL24/Eotaxin-2/MPIF-2 Monoclonal specifically detects CCL24/Eotaxin-2/MPIF-2 in Human samples. It is validated for Western Blot, ELISA.Specifications
CCL24/Eotaxin-2/MPIF-2 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CCL24 | |
Purified recombinant human eotaxin 2, expressed in E. coli as immunogen. | |
0.1 mL | |
Biologically Active Proteins, Cytokine Research | |
6369 | |
Human | |
Purified |
Western Blot, ELISA | |
61016.11 | |
Western Blot, ELISA | |
chemokine (C-C motif) ligand 24, Ckb-6, Eosinophil chemotactic protein 2, Eotaxin-2, member 24, MPIF2Myeloid progenitor inhibitory factor 2, MPIF-2Small-inducible cytokine A24, SCYA24CK-beta-6 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
When used as a capture in human eotaxin 2 sandwich ELISAs, this exhibits less than 0.03 % cross-reactivity with recombinant human eotaxin, MIP1 alpha, and MCP3. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction