Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL27/CTACK Antibody (RM0178-6M35), Alexa Fluor™ 700, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212113AF700
Description
CCL27/CTACK Monoclonal specifically detects CCL27/CTACK in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CCL27/CTACK | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CCL27 | |
Mouse recombinant CCL27/CTACK | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
RM0178-6M35 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
ALP, C-C motif chemokine 27, chemokine (C-C motif) ligand 27, CTACKSmall-inducible cytokine A27, CTAK, cutaneous T-cell attracting chemokine, Cutaneous T-cell-attracting chemokine, ESKINE, IL-11 Ralpha-locus chemokine, ILCCC chemokine ILC, PESKY, SCYA27IL-11 R-alpha-locus chemokine, skinkine, small inducible cytokine subfamily A (Cys-Cys), member 27 | |
Rat | |
Protein A or G purified | |
Biologically Active Proteins | |
10850 | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction