Learn More
Invitrogen™ CCL27 Monoclonal Antibody (124308)

Description
Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
Specifications
Specifications
| Antigen | CCL27 |
| Applications | Immunohistochemistry (Paraffin), Neutralization, Western Blot |
| Classification | Monoclonal |
| Clone | 124308 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | CCL27 |
| Gene Accession No. | Q9Y4X3 |
| Gene Alias | ALP; AW558992; CC chemokine ILC; C-C motif chemokine 27; C-C motif chemokine ligand 27; Ccl27; Ccl27a; chemokine (C-C motif) ligand 27; chemokine (C-C motif) ligand 27A; CTACK; CTAK; cutaneous T-cell attracting chemokine; cutaneous T-cell-attracting chemokine; ESkine; IL-11 R-alpha-locus chemokine; IL-11 Ralpha-locus chemokine; ILC; PESKY; SCYA27; Scya27a; Skinkine; skinskine; small inducible cytokine A27; small inducible cytokine A27a; small inducible cytokine subfamily A (Cys-Cys), member 27; small-inducible cytokine A27 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.