Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCL27 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584941
Description
Immunogen sequence: CTQLYRKPLS DKLLRKVIQV ELQEADGDCH LQAFVLHLAQ RSICIHPQNP SLSQWFEHQE RKLHGTLPKL NFGMLR.
CCL27 gene is one of several CC cytokine genes clustered on the p-arm of chromosome 9. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The protein encoded by this gene is chemotactic for skin-associated memory T lymphocytes. This cytokine may also play a role in mediating homing of lymphocytes to cutaneous sites. It specifically binds to chemokine receptor 10 (CCR10). Studies of a similar murine protein indicate that these protein-receptor interactions have a pivotal role in T cell-mediated skin inflammation.
Specifications
CCL27 | |
Polyclonal | |
Unconjugated | |
CCL27 | |
ALP; AW558992; CC chemokine ILC; C-C motif chemokine 27; C-C motif chemokine ligand 27; Ccl27; Ccl27a; chemokine (C-C motif) ligand 27; chemokine (C-C motif) ligand 27A; CTACK; CTAK; cutaneous T-cell attracting chemokine; cutaneous T-cell-attracting chemokine; ESkine; IL-11 R-alpha-locus chemokine; IL-11 Ralpha-locus chemokine; ILC; PESKY; SCYA27; Scya27a; Skinkine; skinskine; small inducible cytokine A27; small inducible cytokine A27a; small inducible cytokine subfamily A (Cys-Cys), member 27; small-inducible cytokine A27 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10850 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y4X3 | |
CCL27 | |
Recombinant protein corresponding to Human CCL27. Recombinant protein control fragment (Product #RP-108159). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction