Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL28 Antibody (012), DyLight 488, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289585G
Description
CCL28 Monoclonal antibody specifically detects CCL28 in Human samples. It is validated for ELISASpecifications
CCL28 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
012 | |
ELISA | |
chemokine (C-C motif) ligand 28, member 28, MGC71902, Protein CCK1 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human CCL28 (NP_683513.1; Ile23-Tyr127). | |
0.1 mL | |
Biologically Active Proteins | |
56477 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction