Learn More
                                    CCL3/CCL4 Antibody (93342), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB2701MFV450
Description
CCL3/CCL4 Monoclonal antibody specifically detects CCL3/CCL4 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
| CCL3/CCL4 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2b | 
| Western Blot, Flow Cytometry, CyTOF | |
| 93342 | |
| Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| C-C motif chemokine;C-C motif chemokine 3;G0S19-1;LD78ALPHA;MIP1A;MIP-1-alpha;SCYA3 | |
| E. coli-derived recombinant human CCL3/MIP-1 alpha , Ala27-Ala92, Accession # P10147 | |
| RUO | |
| 6348 | |
| Store at 4C in the dark. | 