Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL3/MIP-1 alpha Antibody (39624), DyLight 594, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB450M
Description
CCL3/MIP-1 alpha Monoclonal antibody specifically detects CCL3/MIP-1 alpha in Mouse samples. It is validated for Western BlotSpecifications
CCL3/MIP-1 alpha | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot | |
39624 | |
Western Blot | |
C-C motif chemokine 3, MIP1-(a), AI323804, chemokine (C-C motif) ligand 3, G0S19-1, LD78alpha, Mip1a, MIP-1alpha, MIP1-alpha, Scya3 | |
E. coli-derived recombinant mouse CCL3/MIP-1 alpha , Ala24-Ala92, Accession # P10855 | |
0.1 mL | |
Biologically Active Proteins, Cancer, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Signal Transduction | |
6348 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction