Learn More
CCL3/MIP-1 alpha Antibody (MM0154-11F16), Alexa Fluor™ 532, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP212118AF532
Catalog No. NP212118A32
Only null left
Add to Cart
Description
Description
CCL3/MIP-1 alpha Monoclonal specifically detects CCL3/MIP-1 alpha in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
CCL3/MIP-1 alpha
Monoclonal
Alexa Fluor 532
50 mM sodium borate with 0.05% sodium azide
CCL3
Human recombinant CCL3/MIP-1 alpha
Primary
Human
IgG1
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
MM0154-11F16
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
C-C motif chemokine 3, MIP1-(a), AI323804, chemokine (C-C motif) ligand 3, G0S19-1, LD78alpha, Mip1a, MIP-1alpha, MIP1-alpha, Scya3
Mouse
0.1 mL
6348
Store at 4°C in the dark.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.