Learn More
Invitrogen™ CCL3 (MIP-1 alpha) Monoclonal Antibody (CR3M), PE, eBioscience™
Mouse Monoclonal Antibody
$375.00
Specifications
Antigen | CCL3 (MIP-1 alpha) |
---|---|
Clone | CR3M |
Concentration | 5 μL/Test |
Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
Applications | Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-112-2448
![]() |
Invitrogen™
12970642 |
100 Tests |
Each of 1 for $375.00
|
![]() |
|||||
Description
Description: This CR3M monoclonal antibody reacts with human CCL3. CCL3, also known as MIP-1 alpha (Macrophage Inflammatory Protein 1 alpha), is a member of the CC- subfamily of chemokines and is most closely related to CCL4, or MIP-1 beta. These proteins play critical roles in the recruitment of leukocytes to the site of inflammation. While both CCL3 and CCL4 will attract monocytes, macrophages, and dendritic cells, CCL3 preferentially attracts CD8+ T cells, while CD4+ T cells are more responsive to CCL4. In addition to its chemotactic functions, CCL3 induces inflammatory cytokine secretion, mast cell degranulation, and NK cell activation. It has also been reported to inhibit hematopoetic stem cell proliferation and may be responsible for the maintenance of these cells in a quiescent state. CCL3 signaling is mediated by the G protein-coupled receptors CCR1, CCR4, and CCR5, which are shared with CCL4 and CCL5 (RANTES). CCR5 is the primary co-receptor for HIV entry, which the virus binds through the gp120 envelope protein. All CCR5 ligands demonstrate potent inhibition of virus entry into the cell, both through steric hindrance of gp120-CCR5 interaction, and ligand-mediated receptor internalization. This CR3M clone is specific to CCL3, with no detectable cross-reactivity to Human CCL4 (MIP-1 beta). Applications Reported: This CR3M antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This CR3M antibody has be...
Both MIP-1alpha and MIP-1beta are structurally and functionally related CC chemokines. They participate in host response to invading bacterial, viral, parasite and fungal pathogens by regulating the trafficking and activation state of selected subgroups of inflammatory cells (e.g. macrophages, lymphocytes and NK cells). While both MIP-1alpha and MIP-1beta exert similar effects on monocytes, their effect on lymphocytes differ; with MIP-1alpha selectively attracting CD8+ lymphocytes, and MIP-1beta selectively attracting CD4+ lymphocytes. Additionally, MIP-1alpha and MIP-1beta have also been shown to be potent chemoattractants for B cells, eosinophils and dendritic cells. Both human and murine MIP-1alpha and MIP-1beta are active on human and murine hematopoietic cells.Specifications
CCL3 (MIP-1 alpha) | |
5 μL/Test | |
Flow Cytometry | |
PE | |
Mouse | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
P10147 | |
6348 | |
Primary | |
Affinity Chromatography |
CR3M | |
4°C, store in dark, DO NOT FREEZE! | |
Monoclonal | |
Liquid | |
IgG1 κ | |
Human | |
Ccl3 | |
AI323804; C-C motif chemokine 3; C-C motif chemokine ligand 3; CCL3; chemokine (C-C motif) ligand 3; G0/G1 switch regulatory protein 19-1; G0S19-1; Heparin-binding chemotaxis protein; H-MIP-1-alpha; L2G25B; LD78ALPHA; LD78-alpha; LD78-alpha(4-69); Macrophage inflammatory protein 1 alpha (Small inducible cytokine A3); macrophage inflammatory protein 1-alpha; macrophage inflammatory protein-1alpha; MIP1 (a); mip1 alpha; MIP-1 alpha; MIP1-(a); MIP1A; MIP-1a; MIP-1alpha; MIP-1-alpha; MIP1-alpha; MIP-1-alpha(4-69); PAT 464.1; RP23-320E6.7; Scya3; SIS-alpha; SIS-beta; small inducible cytokine A3; small inducible cytokine A3 (homologous to mouse Mip-1a); small-inducible cytokine A3; Tonsillar lymphocyte LD78 alpha protein; TY-5 | |
Ccl3 | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.