Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL3/MIP Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984740
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4740 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4740 Supplier Bioss Supplier No. BS2452R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Macrophage inflammatory protein-1a (MIP-1a) is an 8kDa member of the C-C chemokine subfamily. C-C chemokines are chemoattractants and activators for monocytes and T cells. MIP-1a has been identified as one of the major HIV-SFs produced by CD8+ T cells, along with MIP-1b and RANTES.

Specifications

Antigen CCL3/MIP
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Ccl3
Gene Accession No. P10147
Gene Alias AI323804; C-C motif chemokine 3; C-C motif chemokine ligand 3; Ccl3; chemokine (C-C motif) ligand 3; G0/G1 switch regulatory protein 19-1; G0S19-1; Heparin-binding chemotaxis protein; H-MIP-1-alpha; L2G25B; LD78ALPHA; LD78-alpha; LD78-alpha(4-69); Macrophage inflammatory protein 1 alpha (Small inducible cytokine A3); Macrophage inflammatory protein 1-alpha; macrophage inflammatory protein-1alpha; MIP1 (a); MIP-1 alpha; MIP1-(a); MIP1A; MIP-1a; MIP-1alpha; MIP-1-alpha; MIP1-alpha; MIP-1-alpha(4-69); PAT 464.1; RP23-320E6.7; SCYA3; SIS-alpha; SIS-beta; small inducible cytokine A3; small inducible cytokine A3 (homologous to mouse Mip-1a); Small-inducible cytokine A3; Tonsillar lymphocyte LD78 alpha protein; TY-5
Gene Symbols Ccl3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from mouse CCL3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6348
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.