Learn More
Invitrogen™ CCL3 Monoclonal Antibody (93342), FITC

Description
CCL3 Monoclonal Antibody for Flow
Specifications
Specifications
| Antigen | CCL3 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 93342 |
| Conjugate | FITC |
| Formulation | Buffered saline solution with ≤0.5% BSA and 0.09% sodium azide |
| Gene | Ccl3 |
| Gene Accession No. | P10147 |
| Gene Alias | AI323804; C-C motif chemokine 3; C-C motif chemokine ligand 3; CCL3; chemokine (C-C motif) ligand 3; G0/G1 switch regulatory protein 19-1; G0S19-1; Heparin-binding chemotaxis protein; H-MIP-1-alpha; L2G25B; LD78ALPHA; LD78-alpha; LD78-alpha(4-69); Macrophage inflammatory protein 1 alpha (Small inducible cytokine A3); macrophage inflammatory protein 1-alpha; macrophage inflammatory protein-1alpha; MIP1 (a); mip1 alpha; MIP-1 alpha; MIP1-(a); MIP1A; MIP-1a; MIP-1alpha; MIP-1-alpha; MIP1-alpha; MIP-1-alpha(4-69); PAT 464.1; RP23-320E6.7; Scya3; SIS-alpha; SIS-beta; small inducible cytokine A3; small inducible cytokine A3 (homologous to mouse Mip-1a); small-inducible cytokine A3; Tonsillar lymphocyte LD78 alpha protein; TY-5 |
| Gene Symbols | Ccl3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.